Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein))

Toimittaja: Biorbyt
BIRBORB12215-100EA 218 EUR
BIRBORB12215-100 BIRBORB12215-500
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR