Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Biorbyt
BIRBORB12225-100EA 0 EUR
BIRBORB12225-100
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR