Anti-V5 Tag E. coli Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB12257-100EA 276 EUR
BIRBORB12257-100 BIRBORB12257-50
Anti-V5 Tag E. coli Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: E. coli
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR