Anti-UMOD Goat Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB12281-100EA 0 EUR
BIRBORB12281-100
Anti-UMOD Goat Polyclonal Antibody
Vasta-aineet
ELISA: 1:128000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: UMOD (uromodulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR