Anti-MCTS1 Goat Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB12294-100EA 0 EUR
BIRBORB12294-100
Anti-MCTS1 Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MCTS1 (malignant T cell amplified sequence 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR