Anti-PROL1 Goat Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB12316-100EA 290 EUR
BIRBORB12316-100
Anti-PROL1 Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PROL1 (proline rich, lacrimal 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR