Anti-SRSF8 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126388-100EA 0 EUR
BIRBORB126388-100 BIRBORB126388-200
Anti-SRSF8 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SRSF8 (serine/arginine-rich splicing factor 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human