Anti-SRY Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126389-200EA 0 EUR
BIRBORB126389-200 BIRBORB126389-100
Anti-SRY Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SRY (sex determining region Y)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human