Anti-STK10 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126394-100EA 0 EUR
BIRBORB126394-100 BIRBORB126394-200
Anti-STK10 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: STK10 (serine/threonine kinase 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human