Anti-STK33 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126395-200EA 0 EUR
BIRBORB126395-200 BIRBORB126395-100
Anti-STK33 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: STK33 (serine/threonine kinase 33)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human