Anti-TAF5L Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126399-200EA 0 EUR
BIRBORB126399-200 BIRBORB126399-100
Anti-TAF5L Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TAF5L (TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human