Anti-TBX22 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126416-200EA 0 EUR
BIRBORB126416-200 BIRBORB126416-100
Anti-TBX22 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TBX22 (T-box 22)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human