Anti-TFAM Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126422-200EA 0 EUR
BIRBORB126422-200 BIRBORB126422-100
Anti-TFAM Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TFAM (transcription factor A, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human