Anti-TFDP1 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126423-200EA 0 EUR
BIRBORB126423-200 BIRBORB126423-100
Anti-TFDP1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TFDP1 (transcription factor Dp-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human