Anti-TIGD1 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126427-200EA 0 EUR
BIRBORB126427-200 BIRBORB126427-100
Anti-TIGD1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TIGD1 (tigger transposable element derived 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human