Anti-TINF2 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126429-100EA 0 EUR
BIRBORB126429-100 BIRBORB126429-200
Anti-TINF2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TINF2 (TERF1 (TRF1)-interacting nuclear factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human