Anti-TP53 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126448-100EA 0 EUR
BIRBORB126448-100 BIRBORB126448-200
Anti-TP53 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human