Anti-TRIM16 Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126457-200EA 0 EUR
BIRBORB126457-200 BIRBORB126457-100
Anti-TRIM16 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TRIM16 (tripartite motif containing 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human