Anti-GCVK_HHV6Z Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126467-200EA 0 EUR
BIRBORB126467-200 BIRBORB126467-100
Anti-GCVK_HHV6Z Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: GCVK_HHV6Z
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human