Anti-UBASH3A Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126471-200EA 0 EUR
BIRBORB126471-200 BIRBORB126471-100
Anti-UBASH3A Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: UBASH3A (ubiquitin associated and SH3 domain containing A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human