Anti-UBE3B Rabbit Polyclonal Antibody
Toimittaja: Biorbyt
BIRBORB126472-200EA 0 EUR
BIRBORB126472-200 BIRBORB126472-100
Anti-UBE3B Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: UBE3B (ubiquitin protein ligase E3B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human