Anti-USP6NL Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
BIRBORB126481-100EA 0 EUR
BIRBORB126481-100 BIRBORB126481-200
Anti-USP6NL Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: USP6NL (USP6 N-terminal like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR