Anti-LEFTY2 Mouse Monoclonal Antibody

Toimittaja: Biorbyt
BIRBORB88273-50EA 0 EUR
BIRBORB88273-50
Anti-LEFTY2 Mouse Monoclonal Antibody
Vasta-aineet
Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: LEFTY2 (left-right determination factor 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR