Anti-ZBTB38 Rabbit polyclonal antibody unconjugated
ABCAAB50664-100EA 771 EUR
ABCAAB50664-100
Anti-ZBTB38 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: ZBTB38 (zinc finger and BTB domain containing 38)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human