Anti-RAF1 Mouse monoclonal antibody unconjugated [clone: RNP1]

ABCAAB50858-100EA 760 EUR
ABCAAB50858-100
Anti-RAF1 Mouse monoclonal antibody unconjugated [clone: RNP1]
Type: Primary
Antigen: RAF1 (v-raf-1 murine leukemia viral oncogene homolog 1)
Clonality: monoclonal
Clone: RNP1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR