Anti-NOL6 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB50875-100EA 743 EUR
ABCAAB50875-100
Anti-NOL6 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: NOL6 (nucleolar protein 6 (RNA-associated))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR