Anti-UBN1 Mouse Monoclonal Antibody [clone: UBN1G12]

Toimittaja: Abcam
ABCAAB50890-100EA 788 EUR
ABCAAB50890-100
Anti-UBN1 Mouse Monoclonal Antibody [clone: UBN1G12]
Vasta-aineet
Type: Primary
Antigen: UBN1 (ubinuclein 1)
Clonality: monoclonal
Clone: UBN1G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR