Anti-VAX2 Mouse Monoclonal Antibody [clone: VAX2A8F12]

Toimittaja: Abcam
ABCAAB50899-100EA 1040 EUR
ABCAAB50899-100
Anti-VAX2 Mouse Monoclonal Antibody [clone: VAX2A8F12]
Vasta-aineet
Type: Primary
Antigen: VAX2 (ventral anterior homeobox 2)
Clonality: monoclonal
Clone: VAX2A8F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR