Anti-TFAP2B Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB50906-100EA 798 EUR
ABCAAB50906-100
Anti-TFAP2B Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TFAP2B (transcription factor AP-2 beta (activating enhancer binding protein 2 beta))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR