Anti-CASP3 Rabbit polyclonal antibody unconjugated

Toimittaja: Abcam
ABCAAB90437-50EA 1050 EUR
ABCAAB90437-50
Anti-CASP3 Rabbit polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR