Anti-PTCH1 Rabbit Polyclonal Antibody
Toimittaja: Abcam
ABCAAB90438-100EA 1030 EUR
ABCAAB90438-100
Anti-PTCH1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PTCH1 (patched 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse