Anti-Neuritin Mouse monoclonal antibody unconjugated [clone: MM0484-1Y36]
ABCAAB90448-50EA 753 EUR
ABCAAB90448-50
Anti-Neuritin Mouse monoclonal antibody unconjugated [clone: MM0484-1Y36]
Type: Primary
Antigen: Neuritin
Clonality: monoclonal
Clone: MM0484-1Y36
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human