Anti-NRTN Mouse monoclonal antibody unconjugated [clone: MM0486-5X51]
ABCAAB90450-50EA 753 EUR
ABCAAB90450-50
Anti-NRTN Mouse monoclonal antibody unconjugated [clone: MM0486-5X51]
Type: Primary
Antigen: NRTN (neurturin)
Clonality: monoclonal
Clone: MM0486-5X51
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human