Anti-PDHA2 Rabbit polyclonal antibody unconjugated
ABCAAB107015-400EA 717 EUR
ABCAAB107015-400
Anti-PDHA2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: PDHA2 (pyruvate dehydrogenase (lipoamide) alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human