Anti-ASS1 Mouse monoclonal antibody unconjugated [clone: 2B10]

Toimittaja: Abcam
ABCAAB124465-100EA 1130 EUR
ABCAAB124465-100
Anti-ASS1 Mouse monoclonal antibody unconjugated [clone: 2B10]
Vasta-aineet
Type: Primary
Antigen: ASS1 (argininosuccinate synthase 1)
Clonality: monoclonal
Clone: 2B10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR