Anti-HAPLN2 Rabbit polyclonal antibody unconjugated

Toimittaja: Abcam
ABCAAB124504-100EA 946 EUR
ABCAAB124504-100
Anti-HAPLN2 Rabbit polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: HAPLN2 (hyaluronan and proteoglycan link protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR