Anti-SLC22A17 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB124506-100EA 950 EUR
ABCAAB124506-100
Anti-SLC22A17 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SLC22A17 (solute carrier family 22, member 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR