Anti-NEDD4L Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB124643-100EA 997 EUR
ABCAAB124643-100
Anti-NEDD4L Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: NEDD4L (neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR