Anti-MUC13 Rabbit Polyclonal Antibody

Toimittaja: Abcam
ABCAAB124654-50EA 938 EUR
ABCAAB124654-50
Anti-MUC13 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MUC13 (mucin 13, cell surface associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR