Anti-HMGB2 Rabbit Monoclonal Antibody [clone: EPR6301]

Toimittaja: Abcam
ABCAAB124670-100EA 1030 EUR
ABCAAB124670-100
Anti-HMGB2 Rabbit Monoclonal Antibody [clone: EPR6301]
Vasta-aineet
Type: Primary
Antigen: HMGB2 (high mobility group box 2)
Clonality: monoclonal
Clone: EPR6301
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR