Anti-MST1R Rabbit Monoclonal Antibody [clone: EPR5598]

Toimittaja: Abcam
ABCAAB124671-100EA 923 EUR
ABCAAB124671-100
Anti-MST1R Rabbit Monoclonal Antibody [clone: EPR5598]
Vasta-aineet
Type: Primary
Antigen: MST1R (macrophage stimulating 1 receptor (c-met-related tyrosine kinase))
Clonality: monoclonal
Clone: EPR5598
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR