Anti-COPS5 Rabbit Monoclonal Antibody [clone: EPR1350]

Toimittaja: Abcam
ABCAAB124720-100EA 935 EUR
ABCAAB124720-100
Anti-COPS5 Rabbit Monoclonal Antibody [clone: EPR1350]
Vasta-aineet
Type: Primary
Antigen: COPS5 (COP9 signalosome subunit 5)
Clonality: monoclonal
Clone: EPR1350
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR