Anti-Interferon Receptor alpha Rabbit Monoclonal Antibody [clone: EPR6244]

Toimittaja: Abcam
ABCAAB124764-100EA 1060 EUR
ABCAAB124764-100
Anti-Interferon Receptor alpha Rabbit Monoclonal Antibody [clone: EPR6244]
Vasta-aineet
Type: Primary
Antigen: Interferon Receptor alpha
Clonality: monoclonal
Clone: EPR6244
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR