Anti-Monoacylglycerol Lipase Rabbit Monoclonal Antibody [clone: EPR6339]

Toimittaja: Abcam
ABCAAB124796-100EA 992 EUR
ABCAAB124796-100
Anti-Monoacylglycerol Lipase Rabbit Monoclonal Antibody [clone: EPR6339]
Vasta-aineet
Type: Primary
Antigen: Monoacylglycerol Lipase
Clonality: monoclonal
Clone: EPR6339
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR