Anti-Niemann Pick C1 Like 1 Rabbit Monoclonal Antibody [clone: EPR5717]

Toimittaja: Abcam
ABCAAB124801-100EA 929 EUR
ABCAAB124801-100
Anti-Niemann Pick C1 Like 1 Rabbit Monoclonal Antibody [clone: EPR5717]
Vasta-aineet
Type: Primary
Antigen: Niemann Pick C1 Like 1
Clonality: monoclonal
Clone: EPR5717
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR