Anti-GSS Rabbit Monoclonal Antibody [clone: EPR6562]

Toimittaja: Abcam
ABCAAB124811-100EA 1030 EUR
ABCAAB124811-100
Anti-GSS Rabbit Monoclonal Antibody [clone: EPR6562]
Vasta-aineet
Type: Primary
Antigen: GSS (glutathione synthetase)
Clonality: monoclonal
Clone: EPR6562
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR