Anti-PI 3 Kinase regulatory subunit 4 Rabbit Monoclonal Antibody [clone: EPR5303]

Toimittaja: Abcam
ABCAAB124817-100EA 989 EUR
ABCAAB124817-100
Anti-PI 3 Kinase regulatory subunit 4 Rabbit Monoclonal Antibody [clone: EPR5303]
Vasta-aineet
Type: Primary
Antigen: PI 3 Kinase regulatory subunit 4
Clonality: monoclonal
Clone: EPR5303
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR