Anti-UGGT Rabbit Monoclonal Antibody [clone: EPR5105]

Toimittaja: Abcam
ABCAAB124879-100EA 933 EUR
ABCAAB124879-100
Anti-UGGT Rabbit Monoclonal Antibody [clone: EPR5105]
Vasta-aineet
Type: Primary
Antigen: UGGT
Clonality: monoclonal
Clone: EPR5105
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR