Anti-EPS8 Rabbit Monoclonal Antibody [clone: EPR6112]

Toimittaja: Abcam
ABCAAB124882-100EA 900 EUR
ABCAAB124882-100
Anti-EPS8 Rabbit Monoclonal Antibody [clone: EPR6112]
Vasta-aineet
Type: Primary
Antigen: EPS8 (epidermal growth factor receptor pathway substrate 8)
Clonality: monoclonal
Clone: EPR6112
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR