Anti-STC1 Rabbit Monoclonal Antibody [clone: EPR6055]

Toimittaja: Abcam
ABCAAB124891-100EA 997 EUR
ABCAAB124891-100
Anti-STC1 Rabbit Monoclonal Antibody [clone: EPR6055]
Vasta-aineet
Type: Primary
Antigen: STC1 (stanniocalcin 1)
Clonality: monoclonal
Clone: EPR6055
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR